EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Acyclic nucleotide analogues based on phosphonic acids].
PMID 9551196
Abstract
The synthesis of novel nucleotide analogues on the basis of ethylphosphonic acid was described. A rigid structural element, an amide or a double bond, was characteristics of the compounds synthesized. The antiviral and cytotoxic activities of these compounds were studied in cell cultures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
289876
Ethylphosphonic acid, 98%
C2H7O3P