EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Towards the optimal antihistamine: studies with ebastine.
PMID 9561405
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9531
Ebastine, ≥98% (HPLC), solid
C32H39NO2