EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
The glutathione dependence of inorganic sulfate formation from L- or D-cysteine in isolated rat hepatocytes.
PMID 9609386
Abstract
The GSH dependence of the metabolic pathways involved in the conversion of cysteine to sulfate in intact cells has been investigated. It was found that hepatocyte-catalysed sulfate formation from added L-cysteine did not occur if hepatocyte GSH was depleted beforehand, but was restored when GSH levels recovered. Furthermore, sulfate formation did not recover in GSH-depleted hepatocytes if GSH synthesis was prevented with buthionine sulfoximine. Thiosulfate formation was, however, markedly enhanced in GSH-depleted hepatocytes. These results suggest that thiosulfate is an intermediate in the formation of inorganic sulfate from L-cysteine and that GSH was required for the conversion of thiosulfate to inorganic sulfate. Much less sulfate was formed if the cysteine was replaced with cysteinesulfinate. Furthermore, sulfate formation from L-cysteine was markedly inhibited by the addition of the transaminase inhibitor DL-cycloserine or the gamma-cystathionase inhibitor DL-propargylglycine. The major routes of sulfate formation from L-cysteine therefore seems to involve pathways that do not involve L-cysteinesulfinate. Similar amounts of sulfate were formed from D-cysteine as L-cysteine. Thiosulfate instead of sulfate was also formed in GSH-depleted hepatocytes. However, sulfate formation from D-cysteine differed from L-cysteine in that it was inhibited by the D-aminoacid oxidase inhibitor sodium benzoate and was not affected by transaminase or gamma-cystathionase inhibitors. These results suggest that thiosulfate is an intermediate in sulfate formation from D-cysteine and involves the oxidation of D-cysteine by D-amino acid oxidase to form beta-mercaptopyruvate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7005
DL-Cycloserine, crystalline
C3H6N2O2
359394
Sodium mercaptopyruvate, 90%, technical grade
C3H3NaO3S