EMAIL THIS PAGE TO A FRIEND
Biochemistry
Role of the colchicine ring A and its methoxy groups in the binding to tubulin and microtubule inhibition.
PMID 9622487
Abstract
The roles of the methoxy substituents on ring A of two ring colchicine (COL) analogues were probed by the synthesis of a number of drugs and the examination of their effect on binding to tubulin, inhibition of microtubule assembly, and induction of GTPase activity. Selective elimination of ring A methoxy groups at positions 2, 3, and 4 weakened all three processes. The effects on binding and inhibition were independent of the nature of ring C (or C'). Specifically, excision of the 2- or 3-methoxy groups weakened binding by ca. 0.4 kcal mol-1, while that of the 4-methoxy group of ring A was weakened by 1.36 +/- 0.15 kcal mol-1. The effect on the inhibition of microtubule assembly, expressed as the equilibrium constant for the binding of the tubulin-drug complex to the end of a microtubule, was more complex and strongly dependent on the nature of ring C (or C'). This was attributed to the abilities of various groups on ring C' to overcome the wobbling in the tubulin-drug complex introduced by the weakening of the anchoring provided by ring A. It is concluded that ring A of COL is not germane to the mechanism of the inhibition of tubulin self-assembly. It serves only as a complex-stabilizing anchor. The control of this process resides in the interactions that key oxygen atoms of ring C of COL or C' of structural analogues establish with the protein. It is proposed that the 4-methoxy group of ring A serves as a key attachment point for immobilization of the drugs on the protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9754
Colchicine, ≥95% (HPLC), powder
C22H25NO6