EMAIL THIS PAGE TO A FRIEND
Genes & development
Inhibition of the anti-apoptotic PI(3)K/Akt/Bad pathway by stress.
PMID 9649498
Abstract
The initiation of apoptosis often transpires in the presence of agents that regulate cell survival. This study evaluated the effects of stress-induced ceramide on the anti-apoptotic activity of the phosphoinositide-3 kinase [PI(3)K] pathway. PI(3)K activity is directly down-regulated by stress-induced ceramide in a dose-dependent manner with rapid kinetics and high specificity. Ceramide inhibition of PI(3)K is dependent on acid-sphingomyelinase. Down-regulation of PI(3)K by ceramide results in inhibition of the kinase Akt and decreased phosphorylation of the death effector Bad. Thus, ceramide levels could act as a general apoptotic rheostat controlling cell survival by regulating PI(3)K anti-apoptotic effector mechanisms.