EMAIL THIS PAGE TO A FRIEND
Regulatory toxicology and pharmacology : RTP
Molinate: rodent reproductive toxicity and its relevance to humans--a review.
PMID 9671566
Abstract
Molinate is a thiocarbamate herbicide used on rice. During the evaluation of the compound for regulatory compliance, an adverse effect on male reproduction in rats was observed. This led to extensive investigations in rats, mice, rabbits, dogs, monkeys, and humans, resulting in a description of the cause of the effect and establishing an empirical case for rodent specificity. More recent investigations have also shown an effect on the ovaries in rodents. A series of investigations into the mechanism of action to support the view that the effect was specific to rodents has been concluded. The results from this investigation are drawn together and show that the mode of action is via a metabolite, the sulfoxide, which is primarily found in rodents, and that the lesion in both rodent sexes is elicited via inhibition of the enzyme neutral cholesterol ester hydrolase, resulting in interference with mobilization of cholesterol from high-density lipoprotein, a path specific to rodents. Thus, the mechanism of action is such that the effect cannot be elicited in other species including humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36171
Molinate, PESTANAL®, analytical standard
C9H17NOS