EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Regulation of rat glutathione S-transferase A5 by cancer chemopreventive agents: mechanisms of inducible resistance to aflatoxin B1.
PMID 9679543
Abstract
The rat can be protected against aflatoxin B1 (AFB1) hepatocarcinogenesis by being fed on a diet containing the synthetic antioxidant ethoxyquin. Evidence suggests that chemoprotection against AFB1 is due to increased detoxification of the mycotoxin by one or more inducible drug-metabolising enzymes. The glutathione S-transferase (GST) isoenzymes in rat liver that contribute to ethoxyquin-induced chemoprotection against AFB1 have been identified by protein purification. This approach resulted in the isolation of several heterodimeric class alpha GST, all of which contained the A5 subunit and possessed at least 50-fold greater activity towards AFB1-8,9-epoxide than previously studied transferases. Molecular cloning and heterologous expression of rat GSTA5-5 has led to the demonstration that it exhibits substantially greater activity for AFB1-8,9-epoxide than other rat transferases. The A5 homodimer can also catalyse the conjugation of glutathione with other epoxides, such as trans-stilbene oxide and 1,2-epoxy-3-(4'-nitrophenoxy)propane, and possesses high catalytic activity for the reactive aldehyde 4-hydroxynonenal. Western blotting has shown that the A5 subunit is not only induced by ethoxyquin but that it is also induced by other cancer chemopreventive agents, such as butylated hydroxyanisole, oltipraz, benzyl isothiocyanate, indole-3-carbinol and coumarin. In addition to GSTA5, we have identified a novel aflatoxin-aldehyde reductase (AFAR) that is similarly induced by ethoxyquin. However, immunoblotting has shown that GSTA5 and AFAR are not always co-ordinately regulated by chemoprotectors. In order to gain a better understanding of the mechanisms responsible for the induction of GSTA5 protein, the GSTA5 gene has been cloned. It was isolated on two overlapping bacteriophage lambda clones and found to be approximately 12 kb in length. The transcriptional start site of GSTA5 has been identified 228 bp upstream from the ATG translational initiation codon. Computer-assisted analysis of the upstream sequence has indicated the presence of a putative antioxidant responsive element (located between -421 and -429 bp) which may be responsible for the induction of GSTA5 by chemopreventive agents.