EMAIL THIS PAGE TO A FRIEND
Clinical and experimental hypertension (New York, N.Y. : 1993)
The cellular mechanism of action of cardiotonic steroids: a new hypothesis.
PMID 9682925
Abstract
Arterial smooth muscle (ASM) contraction is triggered by agonist-evoked Ca2+ mobilization from sarcoplasmic reticulum (SR). The amount of Ca2+ released, and thus, the magnitude of the contractions, depends directly on SR Ca2+ content. Na+ pump inhibition by cardiotonic steroids (CTS) indirectly increases the Ca2+ content of the SR and, thus, contractility. This sequence of events does not, however, account for the multiple Na+ pump alpha subunit isoforms with different affinities for Na+ and for CTS, nor does it explain the cardiotonic and vasotonic effects of low doses of CTS that do not elevate cytosolic Na+ or Ca2+. We show that the Na+ pump high ouabain affinity (alpha3) isoform and the plasmalemmal (PM) Na/Ca exchanger are confined to PM domains that overlie junctional SR in ASM, while low ouabain affinity alpha1 and the PM Ca2+ pump are uniformly distributed in the PM. Thus, low doses of CTS, including an endogenous ouabain-like compound, influence cytosolic Na+ and (indirectly) Ca2+ concentrations only in the cytoplasmic clefts between the PM and junctional SR (a functional unit we call the "plasmerosome"). In turn, this modulates the Ca2+ content of the junctional SR and cell responsiveness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3125
Ouabain octahydrate, ≥95% (HPLC), powder
C29H44O12 · 8H2O