EMAIL THIS PAGE TO A FRIEND
Journal of clinical laser medicine & surgery
Application of the holmium:YAG laser for prostatectomy.
PMID 9728126
Abstract
The authors review the current knowledge regarding the application of the Holmium: YAG laser for prostatectomy. Conventional surgical therapies for benign prostatic hyperplasia (BPH) are effective but associated with relatively high morbidity. Laser prostatectomy, using either Neodymium:YAG or potassium-titanyl-phosphate lasers, has emerged as a new and much safer operative approach to relieve symptoms of benign prostatic hyperplasia. However, these laser wavelengths possess key disadvantages that have limited their acceptability and dissemination in everyday urologic practice. THE authors review their own extensive experience in the development of clinical application of Holmium: YAG laser technology for prostatectomy, as well as the published reports in the current medical literature now dealing with this subject. In multiple clinical trials, Holmium:YAG laser resection of the prostate has proven efficacious in relieving symptomatic BPH. Both objective urodynamic measures of voiding outcomes and symptomatic improvement have been shown to be equivalent to standard electrocautery resection of the prostate. At the same time, these studies have demonstrated the superior safety and hemostasis of Holmium:YAG laser prostatectomy compared to electrocautery resection, similar to prior laser prostatectomy procedure. Unlike prior forms of laser prostatectomy, Holmium:YAG laser resection of the prostate acutely removes all obstructing prostate tissue, so that the postoperative catheterization requirement is typically only overnight and improvement in voiding is immediate. Current operative techniques and the latest technological developments to facilitate Holmium:YAG laser prostatectomy are described. Holmium: YAG laser prostatectomy combines the best features of prior laser prostatectomy technologies, including minimal complications and morbidity, with the efficacy and immediacy of voiding outcomes associated with conventional electrocautery resection of the prostate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
457957
Holmium, chips, 99.9% trace metals basis
Ho
693693
Holmium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Ho
755974
Holmium, dendritic pieces, purified by distillation, 99.99% trace rare earth metals basis
Ho
GF92551259
Holmium, rod, 50mm, diameter 3.8mm, 99%
Ho
GF12230154
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Ho
GF39809863
Holmium, wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF22997188
Holmium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ho
GF59427513
Holmium, wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF74648223
Holmium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Ho
GF88732693
Holmium, wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF13376785
Holmium, rod, 200mm, diameter 3.8mm, 99%
Ho
GF28769541
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Ho
GF68907011
Holmium, rod, 100mm, diameter 3.8mm, 99%
Ho
GF31028543
Holmium, powder, 5g, max. particle size 500 micron, 99.9%
Ho
GF78567654
Holmium, powder, 10g, max. particle size 500 micron, 99.9%
Ho
GF29991673
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ho
GF26625067
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Ho
GF42613006
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Ho
GF60938635
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Ho
GF34764666
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Ho
GF78609640
Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Ho
GF49265253
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Ho
GF07383977
Holmium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ho
GF85502640
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Ho
GF19613684
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Ho
GF38519376
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Ho