EMAIL THIS PAGE TO A FRIEND
Cytogenetics and cell genetics
Assignment of human fatty-acid-coenzyme A ligase, very long-chain 1 gene (FACVL1) to human chromosome band 15q21.2 by fluorescence in situ hybridization.
PMID 9730624