EMAIL THIS PAGE TO A FRIEND
Biochemistry
A novel high-affinity inhibitor for inward-rectifier K+ channels.
PMID 9748337
Abstract
Inward-rectifier K+ channels are a group of highly specialized K+ channels that accomplish a variety of important biological tasks. Inward-rectifier K+ channels differ from voltage-activated K+ channels not only functionally but also structurally. Each of the four subunits of the inward-rectifier K+ channels has only two instead of six transmembrane segments compared to the voltage-activated K+ channels. Thus far, there are no high-affinity ligands that directly target any inward-rectifier K+ channel. In the present study, we identified, purified, and synthesized a protein inhibitor of the inward-rectifier K+ channels. The inhibitor, called tertiapin, blocks a G-protein-gated channel (GIRK1/4) and the ROMK1 channel with nanomolar affinities, but a closely related channel, IRK1, is insensitive to tertiapin. Mutagenesis studies show that teritapin inhibits the channel by binding to the external end of the ion conduction pore.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8316 Tertiapin, >95%
C106H176N34O23S5
T1567 Tertiapin-Q trifluoroacetate salt, ≥97% (HPLC)
C106H175N35O24S4