EMAIL THIS PAGE TO A FRIEND
FEBS letters
Agonist stimulation of B1 and B2 kinin receptors causes activation of the MAP kinase signaling pathway, resulting in the translocation of AP-1 in HEK 293 cells.
PMID 9755866
Abstract
In response to bradykinin, phosphorylated MAP kinases (ERK-1 and ERK-2) were abundantly increased in HEK 293 cells, which overexpress the rat B2 kinin receptor. In a similar way des-Arg9-bradykinin stimulation of B1 kinin receptor-overexpressing HEK 293 cells caused activation of the same species of MAP kinase. Furthermore, nuclear translocation of transcription factor AP-1 was also found in the cells after stimulation with either agonist. PD98059, a MAP kinase kinase (MEK-1) inhibitor, blocked the agonist-induced AP-1 translocation as well as the phosphorylation of the MAP kinases. This communication provides the first evidence for both B1 and B2 kinin receptors mediating the MAP kinase signaling pathway to activate AP-1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4397
Bradykinin Fragment 1-8 acetate salt hydrate, ≥97% (HPLC)
C44H61N11O10 · xC2H4O2 · yH2O