EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
The pyrroloquinoline quinone (PQQ)-containing quinoprotein dehydrogenases.
PMID 9765889
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64682
Pyrroloquinoline quinone, ≥97.0% (HPLC)
C14H6N2O8