EMAIL THIS PAGE TO A FRIEND
Experimental cell research
24R,25-dihydroxyvitamin D3 increases cyclic GMP contents, leading to an enhancement of osteocalcin synthesis by 1,25-dihydroxyvitamin D3 in cultured human osteoblastic cells.
PMID 9770350
Abstract
The effect of the physiological vitamin D metabolite 24R, 25-dihydroxyvitamin D3 [24R,25(OH)2D3] on human osteoblastic cells was assessed. Physiological concentrations (10(-9)-10(-8) M) of 24R, 25(OH)2D3 significantly increased the cyclic guanosine 5'-monophosphate (cGMP) content in the human osteoblastic cells by approximately 200% in 5 to 15 min. In contrast, 24S, 25-dihydroxyvitamin D3 had only a weak effect on the cGMP content, and 1,25-dihydroxyvitamin D3 [1,25(OH)2D3] did not affect the content. The production of osteocalcin was not induced by 10(-9)-10(-8) M of 24R,25(OH)2D3 in the absence of 1,25(OH)2D3. However, the same concentration of 24R,25(OH)2D3 showed stimulatory effects on osteocalcin synthesis in the presence of 10(-9) M 1, 25(OH)2D3. Rp-8Br-cyclic GMP, a specific inhibitor of cyclic GMP-dependent protein kinase, significantly inhibited the cooperative effect of 24R,25(OH)2D3 with 1,25(OH)2D3 on the osteocalcin synthesis, although Rp-8Br-cyclic AMP, a specific inhibitor of cyclic AMP-dependent protein kinase, did not affect the cooperative effect. In addition, okadaic acid enhanced the osteocalcin synthesis induced by 1,25(OH)2D3. These observations suggest that 24R,25(OH)2D3 has a unique activity of increasing cGMP contents in osteoblastic cells, and that the increase in cGMP contents may lead to the cooperative effect of 24R,25(OH)2D3 with 1, 25(OH)2D3 on osteocalcin synthesis. These data support the hypothesis that 24R,25(OH)2D3 has a physiological role in human bone and mineral metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17943
(24R)-24,25-Dihydroxyvitamin D3, ≥98% (vitamin + pre-vitamin, HPLC)
C27H44O3