EMAIL THIS PAGE TO A FRIEND
Immunopharmacology
Triptolide induces apoptotic death of T lymphocyte.
PMID 9826028
Abstract
Extract of Tripterygium wilfordii Hook. f (TWHf) has immunosuppressive activity and has been used as anti-inflammatory agent in traditional Chinese medicine for centuries. Recent studies have demonstrated that triptolide is the major active component in the extract that inhibits antigen or mitogen-induced T cell proliferation. In attempting to investigate its effect on activation of T lymphocytes, we found triptolide induces apoptotic death of T cell hybridomas and peripheral T cells but not that of thymocytes. The triptolide-induced apoptosis is accompanied by increase of DEVD-cleavable caspases activity and degradation of caspase substrate poly (ADP-ribose) polymerase (PARP). A specific inhibitor of caspases, zVAD-FMK, prevents triptolide-induced PARP degradation and DNA fragmentation but not growth arrest. Furthermore, enforced expression of Bcl-2 inhibited triptolide-induced degradation of PARP and apoptosis. These results indicate that triptolide induces T cell apoptosis through activating caspases, and suggest the growth arrest and apoptotic effect of triptolide may contribute to the immunosuppressive activity of TWHf extract.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3652
Triptolide, from Tripterygium wilfordii, ≥98% (HPLC), solid
C20H24O6