EMAIL THIS PAGE TO A FRIEND
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme
[Improvement of polyacrylamide gel media for electrophoresis: enhancement of elasticity by polymer blending].
PMID 9868355