EMAIL THIS PAGE TO A FRIEND
Cytogenetics and cell genetics
Assignment1 of the mitochondrial translational initiation factor 2 gene (MTIF2) to human chromosome 2 bands p16-->p14 by in situ hybridization and with somatic cell hybrids.
PMID 9925935