144592 Aldrich

2-Trichloromethyl-4(3H)-quinazolinone

99%

DOWNLOAD MSDS (PDF)
  • CAS Number 5558-95-2

  • Empirical Formula (Hill Notation) C9H5Cl3N2O

  • Molecular Weight 263.51

  •  EC Number 226-927-2

  •  MDL number MFCD00006886

  •  PubChem Substance ID 24848670

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Building Blocks, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks, Quinazolines More...
assay   99%
mp   211-213 °C(lit.)

Description

Packaging

25 g in poly bottle

Price and Availability

Bulk Order Request


Bulk Order RequestWant a larger quantity than what is offered above? Fill out our new, simplified bulk
order request. Click below to get started.





Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?