CAS Number 62638-00-0
Linear Formula C6H11(CH2)3CO2Li
Molecular Weight 176.18
EC Number 263-664-2
MDL number MFCD00036394
PubChem Substance ID 24853679
Popular Documents: FTIR (PDF)
| Related Categories | Lithium Salts, Materials Science, Metal and Ceramic Science, Salts |
| form | powder and chunks |
| mp | 211-219 °C(lit.) |
Lithium standard salt for organic systems. Lot specific assay on label.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?