CAS Number 110499-97-3
Linear Formula CH3(CH2)5C6H4C6H4CH2CH2C6H4NCS
Molecular Weight 399.59
MDL number MFCD00134447
PubChem Substance ID 24863214
Popular Documents: FTNMR (PDF)
| Related Categories | LC Mesogen, Liquid Crystals, Materials Science, Organic and Printed Electronics |
| assay | 99% |
250 mg in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?