Synonym:
Poly(2,6-di-tert-
CAS Number 107054-29-5
MDL number MFCD00134463
Popular Documents: Specification Sheet (PDF)
| Related Categories | Bases, Chemical Synthesis, Organic Bases, Solid Supported Synthesis, Specialty Synthesis, |
| crosslinking | 1 % cross-linked with divinylbenzene |
| extent of labeling | ~1.8 mmol/g loading |
| particle size | 200-400 mesh |
5, 25 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?