EMAIL THIS PAGE TO A FRIEND
390747 Aldrich

1-Nitropyrazole

98%

  • CAS Number 7119-95-1

  • Empirical Formula (Hill Notation) C3H3N3O2

  • Molecular Weight 113.07

  •  MDL number MFCD00015894

  •  PubChem Substance ID 24864343

Purchase

Properties

Related Categories Building Blocks, C3 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazoles More...
assay   98%
mp   92-94 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability


Search for Answers, Not Just Materials
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?