CAS Number 87188-50-9
Linear Formula HCOC6H4OCO2C(CH3)3
Molecular Weight 222.24
MDL number MFCD00191880
PubChem Substance ID 24864692
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Aldehydes, Building Blocks, C10-C12, Carbonates, Carbonyl Compounds, |
| assay | 97% |
| mp | 71-74 °C(lit.) |
1, 5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?