CAS Number 19236-15-8
Linear Formula Nd(OCH(CH3)2)3
Molecular Weight 321.50
MDL number MFCD00058819
PubChem Substance ID 24865690
Popular Documents: Specification Sheet (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Micro/NanoElectronics, Neodymium, |
| assay | 99.8% trace metals basis |
| form | solid |
| mp | >300 °C(lit.) |
1 g in ampule
Precursor for a catalyst that is used in the living polymerization and block copolymerization of ε-caprolactone.1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?