Synonym:
Fmoc-3-(2-pyridyl)-
CAS Number 185379-39-9
Empirical Formula (Hill Notation) C23H20N2O4
Molecular Weight 388.42
Beilstein Registry Number 8721547
MDL number MFCD00672565
PubChem Substance ID 24870845
| Related Categories | Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acid Derivatives More... |
| assay | ≥98.0% (HPLC) |
| storage temp. | 2-8°C |
1 g in poly bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?