Synonym: Poly(perfluoropropylene oxide)
CAS Number 25038-02-2
Linear Formula HO[CF(CF3)CF2O]nH
MDL number MFCD00804498
PubChem Substance ID 24872934
| vapor pressure | 4 × 10-7 ( 20 °C) |
| refractive index |
n |
| viscosity | 7 cSt(lit.) |
| transition temp | pour point <−54 °C |
| density | 1.86 g/mL at 25 °C |
General purpose oil.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?