CAS Number 312624-17-2
Linear Formula C6H5C6H4CH2ZnBr
Molecular Weight 312.52
MDL number MFCD01311439
PubChem Substance ID 24873131
| concentration | 0.5 M in THF |
| density | 0.983 g/mL at 25 °C |
Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?