Synonym: DABCO® chloride resin, TED-Cl resin
MDL number MFCD04974591
PubChem Substance ID 24880917
| Related Categories | Bases, Chemical Biology, Chemical Synthesis, Conjugation Chemistry: Azides, Alkynes and Other Reagents, Organic Bases, |
| crosslinking | 1 % cross-linked with divinylbenzene |
| extent of labeling | ~2.0 mmol/g N loading |
| particle size | 100-200 mesh |
Catalyst for the epoxidation of alpha, beta-enones with 30% aqueous H2O2 and LiOH.1
5, 25 g in glass bottle
DABCO is a registered trademark of Air Products & Chemicals, Inc.
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?