CAS Number 55956-02-0
Linear Formula (CD3)2CDCl
Molecular Weight 85.58
MDL number MFCD00142901
PubChem Substance ID 24881203
| Related Categories | Alphabetical Listings, C, Stable Isotopes More... |
| isotopic purity | 98 atom % D |
| mol wt | mol wt 85.44 by atom % calculation |
| bp | 34-36 °C(lit.) |
| density | 0.934 g/mL at 25 °C |
| mass shift | M+7 |
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?