Synonym:
(1-Methyl-1H-
CAS Number 179055-21-1
Empirical Formula (Hill Notation) C16H32N2Sn
Molecular Weight 371.15
MDL number MFCD15143648
| Related Categories | Building Blocks, C12 to C37, Chemical Synthesis, Heterocyclic Building Blocks, Organometallic Reagents, |
| assay | 97% |
| refractive index |
n |
| bp | 126-128 °C |
| density | 1.135 g/mL at 25 °C |
1 g in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?