CAS Number 617-35-6
Linear Formula CH3COCOOC2H5
Molecular Weight 116.12
Beilstein Registry Number 1071466
Council of Europe no. 430
EC Number 210-511-2
FEMA Number 2457
Flavis number 9.442
MDL number MFCD00009123
PubChem Substance ID 24901103
Popular Documents: Specification Sheet (PDF) | FTNMR (PDF)
| Related Categories | Alphabetical Listings, E-F, Flavors and Fragrances, Halal Certified Products |
| grade | Halal |
| NI | |
| reg. compliance | FDA 21 CFR (172.515) |
| EU Regulation 1334/2008 & 178/2002 | |
| assay | ≥97% |
| refractive index |
n |
| bp | 144 °C(lit.) |
1 kg in glass bottle
1 sample in glass bottle
10, 25 kg in comp drum
250 g in glass bottle
Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?