| Related Categories | Gas Cylinders and Accessories, Gas Equipment Product Guide, Labware More... |
| material | brass valve |
| feature |
needle valve |
Convenient fully assembled lines for removing liquid from Kilo-Lab cylinders. Line consists of a brass CGA 510 fitting connected to a 48in. length of metal braided PTFE tubing connected to a metal ball valve with metal needle connector. "T" is placed in-line and is equipped with a needle valve and 6mm I.D. septum inlet for nitrogen flush of transfer line.
Kilo-Lab is a registered trademark of Sigma-Aldrich Co. LLC