| Related Categories | Aldrich, Aldrich Safety Equipment, Labware, Manufacturer Browser, Pillows and Socks, |
| W × L | 24 in. × 100 ft |
| color | yellow |
Spilfyter® products are made from a specially formulated, lint-free, 100% PP fabric that is virtually inert and will absorb any liquid with which they come in contact. They are able to collect up to 10 times their own weight in waste material (depending upon its viscosity) in 30s. Saturated Spilfyter® products can be landfilled, neutralized, or incinerated in accordance with federal, state and local regulations.
Spilfyter is a registered trademark of National Packaging Services Corp.