| Related Categories | Chemistry Equipment and Labware, Clamps and Supports, Labware, Tubing |
| diam. | 7/8-1 in. |
Non-corroding, lightweight clamps secure tubing to transfer needles, fittings, etc. Press clamp jaws together with pliers to secure.