Z277444 Aldrich

Whatman® GD/X syringe filters

polypropylene membrane, pore size 0.45 μm, diam. 25 mm

DOWNLOAD MSDS (PDF)

Synonym: Whatman filter, syringe filter, syringe filters

Properties

Related Categories Analytical/Chromatography, Filtration Supplies, GD/X syringe filters, HPLC, HPLC Accessories,
material   polypropylene housing
  polypropylene membrane
sterility   non-sterile
feature   glass fiber prefilter
mfr. no.   Whatman Article No., 28420522 (US reference)
  Whatman, 6878-2504
packaging   pkg of 150 ea
diam.   25 mm
volume   ≤25 mL
pore size   0.45 μm
fitting   inlet female Luer (lock)
  outlet male Luer

Description

General description

The GD/X filter contains a unique prefiltration stack of 100% borosilicate glass. The prefilter will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. The volume of the sample filtered is 3-7 times greater than an unprotected membrane. PP housing. Syringe filters can be autoclaved.

Membranes:

Cellulose Acetate:
for biological solutions, high-loading capacity, and low-protein-binding
Nylon: for aqueous and aqueous organics
Polyethersulfone: for serum, plasma, and tissue culture solutions
Polypropylene: high flow for aqueous and organics
Polysulfone: low-protein-adsorption for serums and plasma
PTFE: chemically inert, will stop aqueous aerosols in gas streams
PVDF: the most suitable membrane for HPLC
Glass microfiber: borosilicate with high-load capacity

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Suitability

Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.

Legal Information

Whatman is a registered trademark of GE Healthcare

Price and Availability

NEW! Ascentis® Express 5 µm
HPLC Columns


New Ascentis Express 5 µm HPLC Columns!Ascentis Express columns, based on 5 µm Fused-Core® particles, offer better performance, faster and more efficient separations, compared to columns based on 3 µm and 5 µm totally porous particles, with no backpressure concerns!


Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?