Z560898 Aldrich

Sigma-Aldrich® safety wash bottles

Water, blue cap, capacity 500 mL

DOWNLOAD MSDS (PDF)

Synonym: labeled bottles, safety bottles, wash bottle

Properties

Related Categories Bottles and Caps, Labware, Wash Bottles More...
material   low-density polyethylene bottle and tube
  blue cap
packaging   pack of 3 ea
capacity   500 mL

Description

Legal Information

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Sigma-Aldrich<SUP>®</SUP> safety wash bottles

Acetone, red cap, capacity 500 mL

Sigma-Aldrich<SUP>®</SUP> safety wash bottles

Ethanol, natural cap, capacity 500 mL

capacity 500 mL (16 oz)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?