Z747041 Aldrich

Whatman® UNIFILTER® plates

96 well, Lysate Clarification, well volume 800 μL, lysate clarification media and 0.45m polypropylene membrane

DOWNLOAD MSDS (PDF)

Synonym: micro plate, multiwell plate, well plates

Properties

Related Categories Labware, Manufacturer Browser, Whatman, Whatman Application Specific Multiwell Plates, Whatman Multiwell Plates,
feature   lysate clarification media and 0.45?m polypropylene membrane
mfr. no.   Whatman Article No., 28420978 (US reference)
  Whatman, 7770-0062
packaging   pkg of 25 ea
well volume   800 μL

Description

Application

The UNIFILTER products are convenient, ready to use, disposable filter bottom microplates. Available in 96 well formats, the UNIFILTER microplates offer a choice of filter media that work well with the most stringent applications. UNIFILTERS can be used to process 96 samples at a time. The unique drip director design allows for analysis of the filtrate or the retentate. UNIFILTERS are available in many different variations of well formats and range from 100 μL to 2 mL, using a wide variety of filtration media.

General description

• Design prevents well to well crosstalk
• Large variety of well volumes economizes on expensive consumables
• Choice of filter media allows control of flow rates and retention characteristics
• Standard overall dimensions fit most plate readers, auto samplers, robots

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

UNIFILTER is a registered trademark of GE Healthcare

Whatman is a registered trademark of GE Healthcare

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?