CAS Number 874-80-6
Empirical Formula (Hill Notation) C9H14BrN
Molecular Weight 216.12
MDL number MFCD00671496
PubChem Substance ID 24845002
| Related Categories | Chemical Synthesis, Ionic Liquids, Pyridinium, Specialty Synthesis |
| assay | ≥99.0% |
| impurities | ≤0.2% water |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?