CAS Number 171877-37-5
Empirical Formula (Hill Notation) C9H9NOS
Molecular Weight 179.24
MDL number MFCD06658220
PubChem Substance ID 24844950
| Related Categories | Asymmetric Synthesis, Chemical Synthesis, Chiral Auxiliaries, Oxazolidinone Derivatives |
| assay | ≥98.0% |
| optical activity | [α]/D -75±5°, c = 0.2 in chloroform |
| optical purity | enantiomeric ratio: 97.0:3.0 (LC) |
A highly selective and efficient chiral auxiliary1 which can be directly reduced to its corresponding aldehyde and the chiral auxiliary by reductive cleavage with diisobutylaluminum hydride.2
Evans-Type Oxazolidinethione And Thiazolidinethione Auxiliaries
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?