02581 Fluka

Methanol solution

for protein sequence analysis, ~50% in H2O

DOWNLOAD MSDS (PDF)
  • CAS Number 67-56-1

  • Empirical Formula (Hill Notation) CH4O

  • Molecular Weight 32.04

  •  Beilstein Registry Number 3662861

  •  MDL number MFCD00004595

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing
grade   for protein sequence analysis
concentration   ~50% in H2O
suitability   in accordance for sequence analysis test

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?