Synonym: Gasoline, Petroleum benzin
CAS Number 8032-32-4
EC Number 232-453-7
MDL number MFCD00081849
Popular Documents: FTNMR (PDF)
| Related Categories | Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Grade, Research Essentials |
| impurities | ≤0.005% sulfur (S) |
| ≤0.02% aromatic content (as toluene) | |
| evapn. residue | ≤0.005% |
| refractive index |
n |
| bp | 150-190 °C |
| density | 0.77 g/mL at 20 °C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?