04710 Fluka

Phenobarbital

purum, ≥99.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: 5-Ethyl-5-phenyl-2,4,6-pyrimidinetrione, 5-Ethyl-5-phenylbarbituric acid, Luminal

  • CAS Number 50-06-6

  • Empirical Formula (Hill Notation) C12H12N2O3

  • Molecular Weight 232.24

  •  Beilstein Registry Number 233363

  •  EC Number 200-007-0

  •  MDL number MFCD00066494

Properties

grade   purum
assay   ≥99.0% (T)
drug control   USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada
mp   174-178 °C

Description

Biochem/physiol Actions

Anesthetic; sedative; hypnotic; anticonvulsant

Other Notes

Sales restrictions may apply

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?