04924 Fluka

Phthalate standard for IC

1.000 g/L in H2O, analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1,2-Benzenedicarboxylic acid, Phthalic acid

  • CAS Number 88-99-3

  • Linear Formula C6H4-1,2-(CO2H)2

  • Molecular Weight 166.13

  •  Beilstein Registry Number 608199

  •  EC Number 231-791-2

  •  MDL number MFCD00002467

  •  PubChem Substance ID 24845783

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Anionic Solutions, Chromatography,
grade   analytical standard
shelf life   (limited shelf life, expiry date on the label)
concentration   1.000 g/L in H2O
mp   210-211 °C (dec.)(lit.)

Description

General description

filtered through a 0.45 μm membrane filter

Preparation Note

prepared with phthalic acid and H2O

Price and Availability

Customers Also Viewed

Bis(2-ethylhexyl) phthalate

PESTANAL®, analytical standard

Dioctyl phthalate

≥99.5%

Phthalic acid

puriss. p.a., ≥99.5% (T)

Phthalic acid

ACS reagent, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?