05334 Fluka

7-Aminoindole

≥95.0% (HPLC)

DOWNLOAD MSDS (PDF)

Synonym: (Indol-7-yl)amine, 7-Indolylamine

  • CAS Number 5192-04-1

  • Empirical Formula (Hill Notation) C8H8N2

  • Molecular Weight 132.16

  •  MDL number MFCD02093960

  •  PubChem Substance ID 24845844

Properties

Related Categories Building Blocks, C7 to C9, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
assay   ≥95.0% (HPLC)
mp   96-100 °C

Description

Caution

may discolor to brown on storage

Application

Reactant for preparation of:
• Fluorescent anion receptors and sensors1,2
• Sulfate receptors3
• Protein Kinase C θ (PKCθ) inhibitors4
• Factor Xa Inhibitors5
• Aurora kinase inhibitor
• Antagonist of Mineralocorticoid Receptor6
• Potent histone deacetylase (HDAC) inhibitors7
• TRPV1 antagonists8
• Derivatives of N-(7-indolyl)benzenesulfonamide as cell cycle inhibitors9

Price and Availability

Customers Also Viewed

4-Aminoindole

97%

5-Aminoindole

97%

6-Aminoindole

97%

7-Nitroindole

97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?