05581 Fluka

Acetylacetone

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 2,4-Pentanedione

  • CAS Number 123-54-6

  • Linear Formula CH3COCH2COCH3

  • Molecular Weight 100.12

  •  Beilstein Registry Number 741937

  •  EC Number 204-634-0

  •  MDL number MFCD00008787

  •  PubChem Substance ID 24845859

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories A-AM, A-B, A-F, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted,
grade   analytical standard
vapor density   3.5 (vs air)
vapor pressure   6 mmHg ( 20 °C)
assay   ≥99.6% (GC)
autoignition temp.   662 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   11.4 %
refractive index   n20/D 1.452(lit.)
  n20/D 1.452
bp   140.4 °C(lit.)
mp   −23 °C(lit.)
density   0.975 g/mL at 25 °C(lit.)
Gene Information   human ... ACHE(43), BCHE(590), CES1(1066)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

100-200 mesh particle size, extent of labeling: 2.5-3.5 mmol/g O loading, 2 % cross-linked with divinylbenzene

Acetylacetone

ReagentPlus®, ≥99%

Acetylacetone

puriss. p.a., ≥99.5% (GC)

Acetylacetone

produced by Wacker, ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?