CAS Number 13031-60-2
Linear Formula NH2(CH2)3COOCH3 · HCl
Molecular Weight 153.61
Beilstein Registry Number 3558996
MDL number MFCD00043270
PubChem Substance ID 24846040
| Related Categories | Chemical Biology, Chemical Synthesis, Others, Peptide Chemistry, Unnatural Amino Acid Derivatives More... |
| assay | ≥99.0% (AT) |
| mp | 120-125 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?