07410 Fluka

Carbohydrate Consumption Broth

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Basal Media for Carbohydrate Utilization,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   beef extract, 1.0 g/L
  bromo cresol purpur, 0.10 g/L
  proteose peptone, 10.0 g/L
  sodium chloride, 5.0 g/L
final pH   6.8±0.2 (25 °C)
suitability   nonselective and differential for Escherichia coli and Coliforms
  nonselective and differential for Listeria
  nonselective and differential for Staphylococcus

Description

Application

Carbohydrate consumption broth is recommended for cultivation and differentiation of Listeria species.

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?