07676 Fluka

Lipase, Thermomyces lanuginosa, CLEA

≥25 U/mg

DOWNLOAD MSDS (PDF)

Synonym: Lipase, Thermomyces lanuginosa, cross-linked enzyme aggregate

Properties

Related Categories Chemical Synthesis, Enzyme-Mediated Synthesis, Hydrolases, Specialty Synthesis
storage temp.   2-8°C

Description

Unit Definition

1 U corresponds to the amount of enzyme which sets free1 μmol butyric acid per minute at pH 7.5 and 40°C (glyceryl tributyrate, Fluka No. 91010, as substrate)

Price and Availability

Customers Also Viewed

solution, ≥100,000 U/g

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?