CAS Number 342573-75-5
Empirical Formula (Hill Notation) C8H16N2O4S
Molecular Weight 236.29
MDL number MFCD06798189
PubChem Substance ID 24846117
| Related Categories | Chemical Synthesis, Imidazolium, Ionic Liquids, Specialty Synthesis |
| grade | for catalysis |
| assay | ≥98.5% (T) |
| impurities | ≤200 ppm water |
| refractive index |
n |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?