CAS Number 2566-90-7
Empirical Formula (Hill Notation) C23H34O2
Molecular Weight 342.51
| Related Categories | Biochemicals and Reagents, Fatty Acids, Lipid Analytical Standards, Lipids, Other Lipid Related Products More... |
| grade | analytical standard |
| assay | ≥96.0% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| concentration | ~100 mg/mL in ethanol |
| storage temp. | −20°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?